1-Chloro-4-cyclohexylbenzene ≥95%

Supplier: AOBChem USA

Warning

47919-1G 47919-25G 47919-500MG 47919-5G
76890-370EA 352.22 USD
76890-370 76890-372 76890-374 76890-376
1-Chloro-4-cyclohexylbenzene ≥95%
1-Chloro-4-cyclohexylbenzene
Formula: C₁₂H₁₅Cl
Storage Temperature: Refrigerator
MDL Number: MFCD11111742
CAS Number: 829-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR