(3-(Benzyloxy)-2-methylphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

47723-1G 47723-25G 47723-500MG 47723-5G
76890-336EA 242.95 USD
76890-336 76890-338 76890-340 76890-342
(3-(Benzyloxy)-2-methylphenyl)methanol ≥95%
(3-(Benzyloxy)-2-methylphenyl)methanol
Formula: C₁₅H₁₆O₂
MW: 228.286 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22041843
CAS Number: 918524-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR