1,3-Dichloro-2,4-difluorobenzene ≥97%

Supplier: AOBChem USA

Warning

47707-1G 47707-25G 47707-5G
76890-326EA 73.21 USD
76890-326 76890-328 76890-330
1,3-Dichloro-2,4-difluorobenzene ≥97%
1,3-Dichloro-2,4-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂Cl₂F₂
MW: 182.98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00142519
CAS Number: 36556-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR