1,2-Dichloro-4-(trifluoromethoxy)benzene ≥95% research grade

Supplier: AOBChem USA

Warning

47534-1G 47534-25G 47534-500MG 47534-5G
76890-310EA 292.81 USD
76890-310 76890-312 76890-314 76890-316
1,2-Dichloro-4-(trifluoromethoxy)benzene ≥95% research grade
1,2-Dichloro-4-(trifluoromethoxy)benzene
Formula: C₇H₃Cl₂F₃O
Storage Temperature: Refrigerator
MDL Number: MFCD22199810
CAS Number: 151276-10-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR