1,3-Dichloro-2-fluoro-5-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

47263-1G 47263-25G 47263-5G
76890-264EA 62.6 USD
76890-264 76890-266 76890-268
1,3-Dichloro-2-fluoro-5-iodobenzene ≥97%
1,3-Dichloro-2-fluoro-5-iodobenzene
Formula: C₆H₂Cl₂FI
Storage Temperature: Refrigerator
MDL Number: MFCD09996900
CAS Number: 133307-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR