1-(2,3-Difluorophenyl)ethanol ≥97% research grade

Supplier: AOBChem USA

Warning

46911-25G 46911-5G
76890-244EA 577.13 USD
76890-244 76890-246
1-(2,3-Difluorophenyl)ethanol ≥97% research grade
1-(2,3-Difluorophenyl)ethanol
Formula: C₈H₈F₂O
MW: 158.145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18911748
CAS Number: 1228690-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR