1-Bromo-2-chloro-5-fluoro-3-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

46645-1G 46645-25G 46645-5G 46645-500MG
76890-210EA 96.55 USD
76890-210 76890-212 76890-214 77646-992
1-Bromo-2-chloro-5-fluoro-3-methoxybenzene ≥97%
1-Bromo-2-chloro-5-fluoro-3-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅BrClFO
MW: 239.469 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34619883
CAS Number: 1783528-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR