1-Bromo-2-(cyclopropylmethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

46466-1G 46466-25G 46466-500MG 46466-5G
76890-194EA 379.81 USD
76890-194 76890-196 76890-198 76890-200
1-Bromo-2-(cyclopropylmethyl)benzene ≥95%
1-Bromo-2-(cyclopropylmethyl)benzene
Formula: C₁₀H₁₁Br
MW: 211.0983 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707177
CAS Number: 1889435-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR