1-(4-Bromo-2-methoxyphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

46412-25G 46412-5G
76890-182EA 342.68 USD
76890-182 76890-184
1-(4-Bromo-2-methoxyphenyl)ethanone ≥97%
1-(4-Bromo-2-methoxyphenyl)ethanone
Formula: C₉H₉BrO₂
MW: 229.07 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11848490
CAS Number: 89368-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR