1-methyl-1H-indole-5-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

46345-1G 46345-25G 46345-5G
76890-168EA 85.94 USD
76890-168 76890-170 76890-172
1-methyl-1H-indole-5-carbaldehyde ≥97%
1-methyl-1H-indole-5-carbaldehyde
Formula: C₁₀H₉NO
MW: 159.188 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08271908
CAS Number: 90923-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR