1-(6-Bromo-2-pyridinyl)-1-ethanol ≥97%

Supplier: AOBChem USA

Warning

46299-1G 46299-25G 46299-500MG 46299-5G
76890-152EA 132.62 USD
76890-152 76890-154 76890-156 76890-158
1-(6-Bromo-2-pyridinyl)-1-ethanol ≥97%
1-(6-Bromo-2-pyridinyl)-1-ethanol
Formula: C₇H₈BrNO
Storage Temperature: Ambient
MDL Number: MFCD12198456
CAS Number: 139163-56-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR