(2-Bromo-3-fluoropyridin-4-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

46229-1G 46229-25G 46229-500MG 46229-5G
76890-138EA 147.47 USD
76890-138 76890-140 76890-142 76890-144
(2-Bromo-3-fluoropyridin-4-yl)methanol ≥97%
(2-Bromo-3-fluoropyridin-4-yl)methanol

Precaución: for Research Use Only

Formula: C₆H₅BrFNO
MW: 206.012 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16606993
CAS Number: 1227601-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR