1-(3-Bromo-2-fluorophenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

46057-1G 46057-25G 46057-5G
76890-100EA 84.88 USD
76890-100 76890-102 76890-104
1-(3-Bromo-2-fluorophenyl)ethanol ≥97%
1-(3-Bromo-2-fluorophenyl)ethanol
Formula: C₈H₈BrFO
Storage Temperature: Ambient
MDL Number: MFCD16140247
CAS Number: 1221715-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR