1-Bromo-4-((4-fluorobenzyl)oxy)benzene ≥97%

Supplier: AOBChem USA

Warning

46025-1G 46025-25G 46025-500MG 46025-5G
76890-084EA 284.33 USD
76890-084 76890-086 76890-088 76890-090
1-Bromo-4-((4-fluorobenzyl)oxy)benzene ≥97%
1-Bromo-4-((4-fluorobenzyl)oxy)benzene
Formula: C₁₃H₁₀BrFO
MW: 281.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05105284
CAS Number: 900911-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR