1-(4-Bromothiazol-2-yl)ethanol ≥97%

Supplier: AOBChem USA

Warning

46007-1G 46007-25G 46007-500MG 46007-5G
76890-070EA 439.22 USD
76890-070 76890-072 76890-074 76890-076
1-(4-Bromothiazol-2-yl)ethanol ≥97%
1-(4-Bromothiazol-2-yl)ethanol
Formula: C₅H₆BrNOS
Storage Temperature: Ambient
MDL Number: MFCD28131428
CAS Number: 1370356-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR