1-(3-Fluoropyridin-4-yl)ethanol ≥95%

Supplier: AOBChem USA

Warning

45970-1G 45970-25G 45970-5G
76890-056EA 62.6 USD
76890-056 76890-058 76890-060
1-(3-Fluoropyridin-4-yl)ethanol ≥95%
1-(3-Fluoropyridin-4-yl)ethanol
Formula: C₇H₈FNO
MW: 141.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18384616
CAS Number: 87674-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR