(3-Bromo-5-fluorophenyl)(morpholino)methanone ≥95%

Supplier: AOBChem USA

Warning

45940-1G 45940-25G 45940-5G
76890-042EA 72.15 USD
76890-042 76890-044 76890-046
(3-Bromo-5-fluorophenyl)(morpholino)methanone ≥95%
(3-Bromo-5-fluorophenyl)(morpholino)methanone
Formula: C₁₁H₁₁BrFNO₂
MW: 288.113 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20046060
CAS Number: 1329457-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR