2-(Benzyloxy)-3-bromo-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

45921-1G 45921-25G 45921-500MG 45921-5G
76890-026EA 290.69 USD
76890-026 76890-028 76890-030 76890-032
2-(Benzyloxy)-3-bromo-5-methylbenzoic acid ≥97%
2-(Benzyloxy)-3-bromo-5-methylbenzoic acid
Formula: C₁₅H₁₃BrO₃
MW: 321.166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707141
CAS Number: 2413441-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR