2-(3-((4-Bromo-2-fluorophenoxy)methyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

45623-1G 45623-25G 45623-500MG 45623-5G
76889-986EA 332.07 USD
76889-986 76889-988 76889-990 76889-992
2-(3-((4-Bromo-2-fluorophenoxy)methyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(3-((4-Bromo-2-fluorophenoxy)methyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₉H₂₁BBrFO₃
Storage Temperature: Ambient
MDL Number: MFCD31916481
CAS Number: 2246736-67-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR