4-Fluoro-2-(trifluoromethyl)benzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-Fluoro-2-(trifluoromethyl)phenyl)methanol

45455-25G 45455-5G
76889-954EA 350.1 USD
76889-954 76889-956
4-Fluoro-2-(trifluoromethyl)benzyl alcohol ≥97%
4-Fluoro-2-(trifluoromethyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₆F₄O
Storage Temperature: Ambient
MDL Number: MFCD00061170
CAS Number: 220227-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR