1-Bromo-4-ethoxy-2-fluoro-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

45381-1G 45381-500MG 45381-5G
76889-940EA 471.04 USD
76889-940 76889-942 76889-944
1-Bromo-4-ethoxy-2-fluoro-5-methylbenzene ≥97%
1-Bromo-4-ethoxy-2-fluoro-5-methylbenzene
Formula: C₉H₁₀BrFO
MW: 233.078 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD34474414
CAS Number: 2384424-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR