(3-Ethyl-5-methylphenyl)boronic acid ≥95% research grade

Supplier: AOBChem USA

Warning

45052-1G 45052-25G 45052-500MG 45052-5G
76889-908EA 338.43 USD
76889-908 76889-910 76889-912 76889-914
(3-Ethyl-5-methylphenyl)boronic acid ≥95% research grade
(3-Ethyl-5-methylphenyl)boronic acid
Formula: C₉H₁₃BO₂
MW: 164.009 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30182380
CAS Number: 2426630-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR