2-(4-Methoxy-3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97% research grade

Supplier: AOBChem USA

Warning

41091-25G 41091-5G
76889-844EA 410.57 USD
76889-844 76889-846
2-(4-Methoxy-3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97% research grade
2-(4-Methoxy-3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₅H₂₃BO₃
MW: 262.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22414439
CAS Number: 568572-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR