[5-(Benzyloxy)-2-methoxyphenyl]boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

40997-1G 40997-25G 40997-500MG 40997-5G
76889-828EA 165.51 USD
76889-828 76889-830 76889-832 76889-834
[5-(Benzyloxy)-2-methoxyphenyl]boronic acid ≥97% research grade
[5-(Benzyloxy)-2-methoxyphenyl]boronic acid
Formula: C₁₄H₁₅BO₄
Storage Temperature: Ambient
MDL Number: MFCD22421493
CAS Number: 2051586-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR