(6-(2,2,2-Trifluoroethoxy)-3-pyridinyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: [6-(2,2,2-Trifluoroethoxy)pyridin-3-yl]boronic acid

40557-1G 40557-25G 40557-500MG 40557-5G
76889-800EA 301.3 USD
76889-800 76889-802 76889-804 76889-806
(6-(2,2,2-Trifluoroethoxy)-3-pyridinyl)boronic acid ≥97%
(6-(2,2,2-Trifluoroethoxy)-3-pyridinyl)boronic acid
Formula: C₇H₇BF₃NO₃
Storage Temperature: Refrigerator
CAS Number: 196083-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR