1-(2-Methoxyethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-[1-(2-Methoxyethyl)-1H-pyrazol-4-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-(2-Methoxyethyl)-1H-pyrazole-4-boronic acid Pinacol ester, 1-(2-Methoxyethyl)-1H-pyrazole-4-boronic acid, pinacol ester

40481-1G 40481-25G 40481-5G
76889-790EA 64.72 USD
76889-790 76889-792 76889-794
1-(2-Methoxyethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole ≥97% research grade
1-(2-Methoxyethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Formula: C₁₂H₂₁BN₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD12405649
CAS Number: 847818-71-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR