1-(2-Fluoro-4-(methylthio)phenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

39757-1G 39757-25G 39757-500MG 39757-5G
76889-774EA 404.21 USD
76889-774 76889-776 76889-778 76889-780
1-(2-Fluoro-4-(methylthio)phenyl)ethanol ≥97%
1-(2-Fluoro-4-(methylthio)phenyl)ethanol
Formula: C₉H₁₁FOS
MW: 186.2464 g/mol
Storage Temperature: Refrigerator
CAS Number: 1781550-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR