2,3-Dimethoxy-5-methylphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (2,3-Dimethoxy-5-methylphenyl)boronic acid

39328-1G 39328-25G 39328-500MG 39328-5G
76889-760EA 299.18 USD
76889-760 76889-762 76889-764 76889-766
2,3-Dimethoxy-5-methylphenylboronic acid ≥95%
2,3-Dimethoxy-5-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BO₄
Storage Temperature: Ambient
MDL Number: MFCD12827469
CAS Number: 396102-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR