4-Benzyloxy-3-fluorophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(4-(Benzyloxy)-3-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

38826-1G 38826-25G 38826-500MG 38826-5G
76889-738EA 233.4 USD
76889-738 76889-740 76889-742 76889-744
4-Benzyloxy-3-fluorophenylboronic acid pinacol ester ≥97%
4-Benzyloxy-3-fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₉H₂₂BFO₃
Storage Temperature: Ambient
MDL Number: MFCD22494101
CAS Number: 1352134-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR