(2-Chloro-4-fluoro-3-methoxyphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

38698-1G 38698-25G 38698-500MG 38698-5G
76889-706EA 292.81 USD
76889-706 76889-708 76889-710 76889-712
(2-Chloro-4-fluoro-3-methoxyphenyl)boronic acid ≥97%
(2-Chloro-4-fluoro-3-methoxyphenyl)boronic acid
Formula: C₇H₇BClFO₃
MW: 204.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23379545
CAS Number: 943831-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR