1,2,3-Trichloro-5-methylbenzene ≥97% research grade

Supplier: AOBChem USA

Warning

38676-1G 38676-500MG 38676-5G
76889-692EA 587.74 USD
76889-692 76889-694 76889-696
1,2,3-Trichloro-5-methylbenzene ≥97% research grade
1,2,3-Trichloro-5-methylbenzene
Formula: C₇H₅Cl₃
Storage Temperature: Ambient
MDL Number: MFCD13689008
CAS Number: 21472-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR