(3-Bromo-5-chloro-2-methoxyphenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

38603-1G 38603-25G 38603-5G 38603-500MG
76889-670EA 96.55 USD
76889-670 76889-672 76889-674 77650-538
(3-Bromo-5-chloro-2-methoxyphenyl)boronic acid ≥95%
(3-Bromo-5-chloro-2-methoxyphenyl)boronic acid
Formula: C₇H₇BBrClO₃
MW: 265.30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19237166
CAS Number: 2096341-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR