2-(2-Methoxyphenoxy)ethanol ≥97%

Supplier: AOBChem USA

Warning

38577-1G 38577-25G 38577-5G
76889-652EA 79.57 USD
76889-652 76889-654 76889-656
2-(2-Methoxyphenoxy)ethanol ≥97%
2-(2-Methoxyphenoxy)ethanol
Formula: C₉H₁₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD01095842
CAS Number: 18181-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR