2-(3-Methoxyphenyl)-1,3,4-oxadiazole ≥97%

Supplier: AOBChem USA

Warning

38558-1G 38558-25G 38558-500MG 38558-5G
76889-636EA 215.37 USD
76889-636 76889-638 76889-640 76889-642
2-(3-Methoxyphenyl)-1,3,4-oxadiazole ≥97%
2-(3-Methoxyphenyl)-1,3,4-oxadiazole

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD09755524
CAS Number: 5378-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR