2-(4-Fluorophenyl)-1,3,4-oxadiazole ≥97% research grade

Supplier: AOBChem USA

Warning

38534-25G 38534-5G
76889-624EA 283.27 USD
76889-624 76889-626
2-(4-Fluorophenyl)-1,3,4-oxadiazole ≥97% research grade
2-(4-Fluorophenyl)-1,3,4-oxadiazole
Formula: C₈H₅FN₂O
MW: 164.137 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11044696
CAS Number: 595567-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR