4-Phenoxybenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-Phenoxyphenyl)methanol

38503-1G 38503-25G 38503-5G
76889-610EA 102.91 USD
76889-610 76889-612 76889-614
4-Phenoxybenzyl alcohol ≥97%
4-Phenoxybenzyl alcohol
Formula: C₁₃H₁₂O₂
MW: 200.237 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01463970
CAS Number: 2215-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR