(5-Chloro-2-(trifluoromethyl)phenyl)(thiomorpholino)methanone ≥97% research grade

Supplier: AOBChem USA

Warning

38488-1G 38488-25G 38488-5G 38488-500MG
76889-598EA 106.09 USD
76889-598 76889-600 76889-602 77646-568
(5-Chloro-2-(trifluoromethyl)phenyl)(thiomorpholino)methanone ≥97% research grade
(5-Chloro-2-(trifluoromethyl)phenyl)(thiomorpholino)methanone
Formula: C₁₂H₁₁ClF₃NOS
Storage Temperature: Refrigerator
MDL Number: MFCD32707086
CAS Number: 2413441-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR