(5-Chloro-2-(trifluoromethyl)phenyl)(pyrrolidin-1-yl)methanone ≥97% research grade

Supplier: AOBChem USA

Warning

38485-1G 38485-25G 38485-5G 38485-500MG
76889-586EA 94.43 USD
76889-586 76889-588 76889-590 77652-748
(5-Chloro-2-(trifluoromethyl)phenyl)(pyrrolidin-1-yl)methanone ≥97% research grade
(5-Chloro-2-(trifluoromethyl)phenyl)(pyrrolidin-1-yl)methanone
Formula: C₁₂H₁₁ClF₃NO
Storage Temperature: Refrigerator
MDL Number: MFCD32707083
CAS Number: 2413441-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR