1-Fluoro-2,3,5-tribromobenzene ≥97%

Supplier: AOBChem USA

Warning

38445-25G 38445-5G
76889-574EA 194.15 USD
76889-574 76889-576
1-Fluoro-2,3,5-tribromobenzene ≥97%
1-Fluoro-2,3,5-tribromobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₃F
Storage Temperature: Ambient
MDL Number: MFCD00142627
CAS Number: 2839-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR