(4-Chloro-2,5-dimethylphenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

38406-1G 38406-25G 38406-500MG 38406-5G
76889-542EA 290.69 USD
76889-542 76889-544 76889-546 76889-548
(4-Chloro-2,5-dimethylphenyl)boronic acid ≥97% research grade
(4-Chloro-2,5-dimethylphenyl)boronic acid
Formula: C₈H₁₀BClO₂
Storage Temperature: Refrigerator
MDL Number: MFCD19671687
CAS Number: 1350512-30-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR