(4-((tert-Butoxycarbonyl)amino)-2-fluorophenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

38278-1G 38278-25G 38278-5G 38278-500MG
76889-482EA 117.76 USD
76889-482 76889-484 76889-486 77652-746
(4-((tert-Butoxycarbonyl)amino)-2-fluorophenyl)boronic acid ≥97%
(4-((tert-Butoxycarbonyl)amino)-2-fluorophenyl)boronic acid
Formula: C₁₁H₁₅BFNO₄
MW: 255.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18250042
CAS Number: 1415960-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR