1-(4-Fluorophenyl)cyclobutan-1-ol ≥95%

Supplier: AOBChem USA

Warning

38226-1G 38226-25G 38226-500MG 38226-5G
76889-414EA 335.25 USD
76889-414 76889-416 76889-418 76889-420
1-(4-Fluorophenyl)cyclobutan-1-ol ≥95%
1-(4-Fluorophenyl)cyclobutan-1-ol
Formula: C₁₀H₁₁FO
MW: 166.192 g/mol
Storage Temperature: Refrigerator
CAS Number: 339365-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR