(4,6-Dibromo-2-fluoro-3-methoxyphenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

38160-1G 38160-25G 38160-500MG 38160-5G
76889-400EA 350.1 USD
76889-400 76889-402 76889-404 76889-406
(4,6-Dibromo-2-fluoro-3-methoxyphenyl)boronic acid ≥97% research grade
(4,6-Dibromo-2-fluoro-3-methoxyphenyl)boronic acid
Formula: C₇H₆BBr₂FO₃
Storage Temperature: Refrigerator
MDL Number: MFCD29917479
CAS Number: 2377606-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR