(E)-ethyl 3-(4-chloro-3,5-dimethylphenyl)acrylate ≥97%

Supplier: AOBChem USA

Warning

38136-1G 38136-25G 38136-500MG 38136-5G
76889-354EA 290.69 USD
76889-354 76889-356 76889-358 76889-360
(E)-ethyl 3-(4-chloro-3,5-dimethylphenyl)acrylate ≥97%
(E)-ethyl 3-(4-chloro-3,5-dimethylphenyl)acrylate
Formula: C₁₃H₁₅ClO₂
MW: 238.71 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32707063
CAS Number: 2206709-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR