2-(4-Chlorophenyl)imidazo[1,2-a]pyrimidine ≥95%

Supplier: AOBChem USA

Warning

38102-1G 38102-25G 38102-500MG 38102-5G
76889-324EA 168.69 USD
76889-324 76889-326 76889-328 76889-330
2-(4-Chlorophenyl)imidazo[1,2-a]pyrimidine ≥95%
2-(4-Chlorophenyl)imidazo[1,2-a]pyrimidine
Formula: C₁₂H₈ClN₃
MW: 229.665 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00453236
CAS Number: 56921-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR