(4-Bromo-2-fluoro-3-methylphenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

38096-1G 38096-25G 38096-5G
76889-310EA 63.66 USD
76889-310 76889-312 76889-314
(4-Bromo-2-fluoro-3-methylphenyl)boronic acid ≥97% research grade
(4-Bromo-2-fluoro-3-methylphenyl)boronic acid
Formula: C₇H₇BBrFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD27935898
CAS Number: 2377605-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR