N-(3-Bromo-5-nitrobenzoyl)morpholine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (3-Bromo-5-nitrophenyl)(morpholino)methanone

38065-25G 38065-5G
76889-278EA 453.01 USD
76889-278 76889-280
N-(3-Bromo-5-nitrobenzoyl)morpholine ≥97%
N-(3-Bromo-5-nitrobenzoyl)morpholine

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrN₂O₄
MW: 315.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09475915
CAS Number: 941294-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR