1-Bromo-2-propoxybenzene ≥97% research grade

Supplier: AOBChem USA

Warning

38060-1G 38060-25G 38060-500MG 38060-5G
76889-266EA 177.18 USD
76889-266 76889-268 76889-270 76889-272
1-Bromo-2-propoxybenzene ≥97% research grade
1-Bromo-2-propoxybenzene
Formula: C₉H₁₁BrO
MW: 215.087 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09937799
CAS Number: 54554-81-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR