(2-Fluoro-3-propoxyphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

38000-25G 38000-5G
76889-258EA 488.02 USD
76889-258 76889-260
(2-Fluoro-3-propoxyphenyl)boronic acid ≥97%
(2-Fluoro-3-propoxyphenyl)boronic acid
Formula: C₉H₁₂BFO₃
MW: 197.999 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19237181
CAS Number: 871126-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR