(3-Bromo-5-chlorophenyl)(piperidin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

37943-1G 37943-25G 37943-500MG 37943-5G
76889-210EA 153.84 USD
76889-210 76889-212 76889-214 76889-216
(3-Bromo-5-chlorophenyl)(piperidin-1-yl)methanone ≥97%
(3-Bromo-5-chlorophenyl)(piperidin-1-yl)methanone
Formula: C₁₂H₁₃BrClNO
MW: 302.5947 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29043944
CAS Number: 1327121-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR